Merck
  • Defining total-body AIDS-virus burden with implications for curative strategies.
Defining total-body AIDS-virus burden with implications for curative strategies.
Nature medicine (2017-10-03)
Jacob D Estes, Cissy Kityo, Francis Ssali, Louise Swainson, Krystelle Nganou Makamdop, Gregory Q Del Prete, Steven G Deeks, Paul A Luciw, Jeffrey G Chipman, Gregory J Beilman, Torfi Hoskuldsson, Alexander Khoruts, Jodi Anderson, Claire Deleage, Jacob Jasurda, Thomas E Schmidt, Michael Hafertepe, Samuel P Callisto, Hope Pearson, Thomas Reimann, Jared Schuster, Jordan Schoephoerster, Peter Southern, Katherine Perkey, Liang Shang, Stephen W Wietgrefe, Courtney V Fletcher, Jeffrey D Lifson, Daniel C Douek, Joseph M McCune, Ashley T Haase, Timothy W Schacker
ABSTRACT
In the quest for a functional cure or the eradication of HIV infection, it is necessary to know the sizes of the reservoirs from which infection rebounds after treatment interruption. Thus, we quantified SIV and HIV tissue burdens in tissues of infected nonhuman primates and lymphoid tissue (LT) biopsies from infected humans. Before antiretroviral therapy (ART), LTs contained >98% of the SIV RNA
Sigma-Aldrich
Fumitremorgin C, from Neosartorya fischeri, film