Merck
  • The Dynamics of mRNA Turnover Revealed by Single-Molecule Imaging in Single Cells.
The Dynamics of mRNA Turnover Revealed by Single-Molecule Imaging in Single Cells.
Molecular cell (2017-10-24)
Ivana Horvathova, Franka Voigt, Anna V Kotrys, Yinxiu Zhan, Caroline G Artus-Revel, Jan Eglinger, Michael B Stadler, Luca Giorgetti, Jeffrey A Chao
ABSTRACT
RNA degradation plays a fundamental role in regulating gene expression. In order to characterize the spatiotemporal dynamics of RNA turnover in single cells, we developed a fluorescent biosensor based on dual-color, single-molecule RNA imaging that allows intact transcripts to be distinguished from stabilized degradation intermediates. Using this method, we measured mRNA decay in single cells and found that individual degradation events occur independently within the cytosol and are not enriched within processing bodies. We show that slicing of an mRNA targeted for endonucleolytic cleavage by the RNA-induced silencing complex can be observed in real time in living cells. This methodology provides a framework for investigating the entire life history of individual mRNAs from birth to death in single cells.
Sigma-Aldrich
MISSION® pLKO.1-puro Luciferase shRNA Control Plasmid DNA, shRNA sequence targeting luciferase
Pricing and availability is not currently available.