Merck
  • Adjusting the catalytic properties of cobalt ferrite nanoparticles by pulsed laser fragmentation in water with defined energy dose.
Adjusting the catalytic properties of cobalt ferrite nanoparticles by pulsed laser fragmentation in water with defined energy dose.
Scientific reports (2017-10-17)
Friedrich Waag, Bilal Gökce, Chakrapani Kalapu, Georg Bendt, Soma Salamon, Joachim Landers, Ulrich Hagemann, Markus Heidelmann, Stephan Schulz, Heiko Wende, Nils Hartmann, Malte Behrens, Stephan Barcikowski
ABSTRACT
Highly active, structurally disordered CoFe
Sigma-Aldrich
Cobalt iron oxide, nanopowder, 30 nm particle size (TEM), 99% trace metals basis
Pricing and availability is not currently available.