Merck
  • Retinoic acid signaling determines the fate of uterine stroma in the mouse Müllerian duct.
Retinoic acid signaling determines the fate of uterine stroma in the mouse Müllerian duct.
Proceedings of the National Academy of Sciences of the United States of America (2016-12-03)
Tadaaki Nakajima, Taisen Iguchi, Tomomi Sato
ABSTRACT
The Müllerian duct develops into the oviduct, uterus, and vagina, all of which are quite distinct in their morphology and function. The epithelial fate of these female reproductive organs in developing mice is determined by factors secreted from the stroma; however, how stromal differentiation occurs in the female reproductive organs derived from the Müllerian duct is still unclear. In the present study, roles of retinoic acid (RA) signaling in developing female reproductive tracts were investigated. Retinol dehydrogenase 10 (RDH10) and aldehyde dehydrogenase family 1 subfamily A2 (ALDH1A2) mRNAs and proteins and transactivation activity of endogenous RA were found in the stroma of proximal Müllerian ducts and gradually decreased from the proximal to caudal regions in fetal mice. In organ-cultured Müllerian ducts, retinaldehyde or RA treatment induced uterine epithelial differentiation, defined as a layer of columnar epithelial cells negative for oviductal and vaginal epithelial markers. In contrast, inhibition of RA receptor (RAR) signaling induced vaginal epithelial differentiation, characterized as vaginal epithelial marker genes-positive stratified epithelium. Grafting experiments of the organ-cultured Müllerian duct revealed irreversible epithelial fate determination. Although RAR did not directly bind to the homeobox A10 (Hoxa10) promoter region, RA-RAR signaling stimulated Hoxa10 expression. Thus, RA-RAR signaling in the Müllerian duct determines the fate of stroma to form the future uterus and vagina.
Sigma-Aldrich
AGN 193109, ≥98% (HPLC)
Pricing and availability is not currently available.