Merck
  • Ionic Circuits Powered by Reverse Electrodialysis for an Ultimate Iontronic System.
Ionic Circuits Powered by Reverse Electrodialysis for an Ultimate Iontronic System.
Scientific reports (2017-10-27)
Seok Hee Han, Seung-Ryong Kwon, Seol Baek, Taek-Dong Chung
ABSTRACT
Despite numerous reports on iontronic devices, there has been no whole circuit working in aqueous media including even power source. Herein, we introduce complete ionic circuits powered by reverse electrodialysis (RED) for the first time without employing any electronic components. The RED-driven polyelectrolyte diode successfully shows rectification behavior which is verified by monitoring dynamic ion distribution through fluorescence in real-time. We can also turn on and off the voltage applied to the circuit, and apply an arbitrary voltage by precisely manipulating the pressure imposed to an elastic connection tube filled with electrolyte. Furthermore, this new concept containing ionic power source advances to a more sophisticated ionic OR logic gate. The proposed system paves the way to develop not only passive iontronic devices (e.g. current ionic diode), but active ones requiring a source of energy, particularly such as a neuron-like information processor powered by fully ionic systems, and thereby aqueous computers.
Sigma-Aldrich
2-Acrylamido-2-methyl-1-propanesulfonic acid, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
2-Hydroxy-4′-(2-hydroxyethoxy)-2-methylpropiophenone, 98%
Pricing and availability is not currently available.