Merck
  • A New Generation of Arachidonic Acid Analogues as Potential Neurological Agent Targeting Cytosolic Phospholipase A
A New Generation of Arachidonic Acid Analogues as Potential Neurological Agent Targeting Cytosolic Phospholipase A
Scientific reports (2017-10-24)
Cheng Yang Ng, Srinivasaraghavan Kannan, Yong Jun Chen, Francis Chee Kuan Tan, Wee Yong Ong, Mei Lin Go, Chandra S Verma, Chian-Ming Low, Yulin Lam
ABSTRACT
Cytosolic phospholipase A
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, γ-irradiated, BioXtra, suitable for cell culture
Pricing and availability is not currently available.