Merck
  • Identification of Novel Hydrogen-Substituted Polyfluoroalkyl Ether Sulfonates in Environmental Matrices near Metal-Plating Facilities.
Identification of Novel Hydrogen-Substituted Polyfluoroalkyl Ether Sulfonates in Environmental Matrices near Metal-Plating Facilities.
Environmental science & technology (2017-09-19)
Yongfeng Lin, Ting Ruan, Aifeng Liu, Guibin Jiang
ABSTRACT
Environmental occurrence and behaviors of 6:2 chlorinated polyfluoroalkyl ether sulfonate (Cl-6:2 PFESA, with trade name F-53B) have been receiving increased attention recently. Nevertheless, its potential fates under diversified conditions remain concealed. In this study, susceptibility of Cl-6:2 PFESA to reductive dehalogenation was tested in an anaerobic super-reduced cyanocobalamin assay. A rapid transformation of dosed Cl-6:2 PFESA was observed, with a hydrogen-substituted polyfluoroalkyl ether sulfonate (1H-6:2 PFESA) identified as the predominant product by a nontarget screening workflow. With the aid of laboratory-purified standards, hydrogen-substituted PFESA analogues (i.e., 1H-6:2 and 1H-8:2 PFESA) were further found in river water and sediment samples collected from two separate regions near metal-plating facilities. Geometric mean concentrations of 560 pg/L (river water) and 11.1 pg/g (sediment) for 1H-6:2 PFESA and 11.0 pg/L (river water) and 7.69 pg/g (sediment) for 1H-8:2 PFESA were measured, and both analytes consisted average compositions of 1% and 0.1% among the 18 monitored per- and polyfluoroalkyl sulfonate and carboxylate pollutants, respectively. To our knowledge, this is the first to report existence of polyfluoroalkyl sulfonates with both hydrogen and ether functional group in the environment.
Sigma-Aldrich
Titanium(III) chloride solution, JIS special grade, ≥20.0%