Merck
  • Hexakis [60]Fullerene Adduct-Mediated Covalent Assembly of Ruthenium Nanoparticles and Their Catalytic Properties.
Hexakis [60]Fullerene Adduct-Mediated Covalent Assembly of Ruthenium Nanoparticles and Their Catalytic Properties.
Chemistry (Weinheim an der Bergstrasse, Germany) (2017-05-26)
Faqiang Leng, Iann C Gerber, Pierre Lecante, Ahmed Bentaleb, Antonio Muñoz, Beatriz M Illescas, Nazario Martín, Georgian Melinte, Ovidiu Ersen, Hervé Martinez, M Rosa Axet, Philippe Serp
ABSTRACT
The C
Sigma-Aldrich
Tetrabromomethane, ReagentPlus®, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Diethyl malonate, ReagentPlus®, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Allyl methyl sulfone, 96%
Pricing and availability is not currently available.