Merck
  • The use of atmospheric pressure plasma as a curing process for canned ground ham.
The use of atmospheric pressure plasma as a curing process for canned ground ham.
Food chemistry (2017-09-28)
Juri Lee, Kyung Jo, Yubong Lim, Hee Joon Jeon, Jun Ho Choe, Cheorun Jo, Samooel Jung
ABSTRACT
This study investigated the potential use of atmospheric pressure plasma (APP) treatment as a curing process for canned ground ham. APP treatment for 60min while mixing increased the nitrite content in the meat batters from 0.64 to 60.50mgkg
Sigma-Aldrich
1,1,3,3-Tetraethoxypropane, ≥96%