Merck
  • Rhodamine-based conjugated polymers: potentiometric, colorimetric and voltammetric sensing of mercury ions in aqueous medium.
Rhodamine-based conjugated polymers: potentiometric, colorimetric and voltammetric sensing of mercury ions in aqueous medium.
The Analyst (2017-08-22)
Rukiye Ayranci, Dilek Odaci Demirkol, Suna Timur, Metin Ak
ABSTRACT
Herein, we report the synthesis and characterization of a new rhodamine-based monomer (RD-CZ), and an investigation of the optical and electrochemical properties of the corresponding polymer (P(RD-CZ)), which was electropolymerized on an ITO electrode. The resulting P(RD-CZ) polymer film was used as a simple and novel multi-signal sensor platform, which demonstrates ion-selective potentiometric, colorimetric and voltammetric responses in aqueous media for the first time. P(RD-CZ) exhibits excellent selectivity for Hg
Sigma-Aldrich
9H-Carbazole-9-ethanol, 95%
Pricing and availability is not currently available.