Merck
  • Deposition of metallic silver coatings by Aerosol Assisted MOCVD using two new silver β-diketonate adduct metalorganic precursors.
Deposition of metallic silver coatings by Aerosol Assisted MOCVD using two new silver β-diketonate adduct metalorganic precursors.
Dalton transactions (Cambridge, England : 2003) (2017-08-05)
H Liu, S Battiato, A L Pellegrino, P Paoli, P Rossi, C Jiménez, G Malandrino, D Muñoz-Rojas
ABSTRACT
This work reports two new silver metalorganic precursors for the chemical vapor deposition of Ag metallic coatings. Both precursors are based on β-diketonate adducts, namely, Ag(hfac)(L) (H-hfac = 1,1,1,5,5,5-hexafluoro-2,4-pentanedione), where L is 1,10-phenanthroline (phen) or 2,5,8,11-tetraoxadodecane (triglyme). Using these ligands, the designed precursors have better solubility in alcoholic solvents and are less toxic and costly than previously reported ones. The new precursors have been characterized and their crystallographic structure solved. With the new triglyme precursor, [Ag(triglyme)
Sigma-Aldrich
1,10-Phenanthroline, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Triethylene glycol dimethyl ether, ReagentPlus®, 99%
Pricing and availability is not currently available.