Merck
Potent and Selective Covalent Quinazoline Inhibitors of KRAS G12C.
Cell chemical biology (2017-08-07)
Mei Zeng, Jia Lu, Lianbo Li, Frederic Feru, Chunshan Quan, Thomas W Gero, Scott B Ficarro, Yuan Xiong, Chiara Ambrogio, Raymond M Paranal, Marco Catalano, Jay Shao, Kwok-Kin Wong, Jarrod A Marto, Eric S Fischer, Pasi A Jänne, David A Scott, Kenneth D Westover, Nathanael S Gray
ABSTRACT
Targeted covalent small molecules have shown promise for cancers driven by KRAS G12C. Allosteric compounds that access an inducible pocket formed by movement of a dynamic structural element in KRAS, switch II, have been reported, but these compounds require further optimization to enable their advancement into clinical development. We demonstrate that covalent quinazoline-based switch II pocket (SIIP) compounds effectively suppress GTP loading of KRAS G12C, MAPK phosphorylation, and the growth of cancer cells harboring G12C. Notably we find that adding an amide substituent to the quinazoline scaffold allows additional interactions with KRAS G12C, and remarkably increases the labeling efficiency, potency, and selectivity of KRAS G12C inhibitors. Structural studies using X-ray crystallography reveal a new conformation of SIIP and key interactions made by substituents located at the quinazoline 2-, 4-, and 7-positions. Optimized lead compounds in the quinazoline series selectively inhibit KRAS G12C-dependent signaling and cancer cell growth at sub-micromolar concentrations.
Sigma-Aldrich
Monoclonal Anti-KRAS antibody produced in mouse, clone 4F3, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
cis-11-Methyl-2-dodecenoic acid, ≥90.0% (HPLC)
Supelco
SYPRO® Orange Protein Gel Stain