Merck
  • Characterisation of the selective binding of antibiotics vancomycin and teicoplanin by the VanS receptor regulating type A vancomycin resistance in the enterococci.
Characterisation of the selective binding of antibiotics vancomycin and teicoplanin by the VanS receptor regulating type A vancomycin resistance in the enterococci.
Biochimica et biophysica acta (2017-05-18)
C S Hughes, E Longo, M K Phillips-Jones, R Hussain
ABSTRACT
A-type resistance towards "last-line" glycopeptide antibiotic vancomycin in the leading hospital acquired infectious agent, the enterococci, is the most common in the UK. Resistance is regulated by the VanR
Sigma-Aldrich
N-Acetylmuramic acid, ≥98% (TLC)
Pricing and availability is not currently available.