Merck
  • Increasing the affinity of cationized polyacrylamide-paclitaxel nanoparticles towards colon cancer cells by a surface recognition peptide.
Increasing the affinity of cationized polyacrylamide-paclitaxel nanoparticles towards colon cancer cells by a surface recognition peptide.
International journal of pharmaceutics (2017-08-29)
Sanjay Tiwari, Boaz Tirosh, Abraham Rubinstein
ABSTRACT
Nanoparticles of cationized polyacrylamide (CPAA NP), decorated with the recognition peptide VRPMPLQ (to produce CPAA-Pep NP), were prepared, characterized and tested biologically. They were designed to target dysplasia regions in the colon, characterized by overexpressed sialic acid. This targetability was augmented by the addition of VRPMPLQ. Their mean hydrodynamic size was 137nm with narrow size distribution and positive zeta potential. When incubated with three types of colon cancer cells, a 10-fold increase in the cell's uptake was found for the CPAA-Pep NP compared with the CPAA NP. The use of a scrambled sequence of the VRPMPLQ peptide and competition studies, employing excess of the free peptide verified the specific nature of the NP cellular uptake. Nanoparticles loaded with paclitaxel with and without VRPMPLQ indicated an improved pro-apoptotic activity of the CPAA-Pep NP. It is speculated that both positive charge and the presence of VRPMPLQ could serve as an improved strategy to deliver nanoparticles loaded with cytotoxic drugs for the treatment of colon cancer.
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide, ≥97.0% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
2-Pentynoic acid, 97%
Pricing and availability is not currently available.