Merck
  • Comparative insights into the saccharification potentials of a relatively unexplored but robust
Comparative insights into the saccharification potentials of a relatively unexplored but robust
Biotechnology for biofuels (2017-03-28)
Funso Emmanuel Ogunmolu, Navya Bhatt Kammachi Jagadeesha, Rakesh Kumar, Pawan Kumar, Dinesh Gupta, Syed Shams Yazdani
ABSTRACT
GH7 cellobiohydrolases (CBH1) are vital for the breakdown of cellulose. We had previously observed the enzyme as the most dominant protein in the active cellulose-hydrolyzing secretome of the hypercellulolytic ascomycete- There were marginal differences observed in the stability of both enzymes, with To the best of our knowledge, this is the first report about a comprehensive and comparative description of CBH1 from hypercellulolytic ascomycete-
Sigma-Aldrich
Cellotetraose, ≥85% (HPLC)