Merck
  • Potential-resolved "in-electrode" type electrochemiluminescence immunoassay based on functionalized g-C
Potential-resolved "in-electrode" type electrochemiluminescence immunoassay based on functionalized g-C
Biosensors & bioelectronics (2017-04-17)
Zhiyong Guo, Lin Wu, Yufang Hu, Sui Wang, Xing Li
ABSTRACT
Here, a novel potential-resolved "in-electrode" type electrochemiluminescence (ECL) immunosensor was fabricated based on two different types of luminant Ru-NH
Sigma-Aldrich
2-(Dibutylamino)ethanol, 99%
Pricing and availability is not currently available.