Merck
Artificial web of disclination lines in nematic liquid crystals.
Nature communications (2017-09-01)
Mengfei Wang, Yannian Li, Hiroshi Yokoyama
ABSTRACT
Disclinations are topological singularities of molecular arrangement in liquid crystals, which typically occur when the average orientation of molecules makes a π rotation along a fictitious closed loop taken inside the liquid crystal. Depending on the sense of molecular rotation, the disclination lines are either of 1/2 or -1/2 strength. When two disclination lines with the opposite strength meet, they are annihilated without trace. It is hence generally considered difficult in the nematic phase to stabilize a condensed array of free-standing disclination lines without the aid of topological objects like colloidal inclusions. Here we show that a free-standing web of 1/2-strength twist disclination lines can be stably formed in thin liquid crystal cells by means of a judicious combination of orientationally patterned confining surfaces fabricated by the micropatterned photoalignment technique. Theoretical model indicates that disclination lines are held apart at the intersection by a repulsive force generated by the Frank elasticity.Disclination lines are topological defects in molecular orientation widely found in liquid crystals. Here Wang et al. use a surface patterning technique to produce a very stable freestanding 3D array of ½ twist disclinations, which could be exploited in a variety of nanometre scale applications.
Sigma-Aldrich
2-Methyl-4′-(methylthio)-2-morpholinopropiophenone, 98%
Pricing and availability is not currently available.