Merck
  • Palladium Nanoparticles Encapsulated in Hollow Titanosilicate Spheres as an Ideal Nanoreactor for One-pot Oxidation.
Palladium Nanoparticles Encapsulated in Hollow Titanosilicate Spheres as an Ideal Nanoreactor for One-pot Oxidation.
Chemistry (Weinheim an der Bergstrasse, Germany) (2016-11-26)
Yasutaka Kuwahara, Takahiro Ando, Hiroto Kango, Hiromi Yamashita
ABSTRACT
One-pot reaction involving Pd-catalyzed H
Sigma-Aldrich
Titanium(IV) propoxide, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, 99%
Pricing and availability is not currently available.