Merck
  • Transcriptome-Wide Identification of Novel UV-B- and Light Modulated Flavonol Pathway Genes Controlled by VviMYBF1.
Transcriptome-Wide Identification of Novel UV-B- and Light Modulated Flavonol Pathway Genes Controlled by VviMYBF1.
Frontiers in plant science (2017-07-12)
Stefan Czemmel, Janine Höll, Rodrigo Loyola, Patricio Arce-Johnson, José Antonio Alcalde, José Tomás Matus, Jochen Bogs
ABSTRACT
Flavonols constitute a group of flavonoids with important photoprotective roles in plants. In addition, flavonol content and composition greatly influences fruit quality. We previously demonstrated that the grapevine R2R3-MYB transcription factor (TF) VviMYBF1 promotes flavonol accumulation by inducing the expression of flavonol synthase (
Sigma-Aldrich
3-Hydroxyflavone, ≥98%
Pricing and availability is not currently available.