Merck
  • Inhibition of polyamine synthesis causes entry of the mouse blastocyst into embryonic diapause.
Inhibition of polyamine synthesis causes entry of the mouse blastocyst into embryonic diapause.
Biology of reproduction (2017-06-24)
Jane C Fenelon, Bruce D Murphy
ABSTRACT
Embryonic diapause is a common reproductive strategy amongst mammals, requiring an intimate cross-talk between the endometrium and the blastocyst. To date, the precise molecular signals responsible are unknown in the mouse or any other mammal. Previous studies in the mink implicate polyamines as major regulators of the control of diapause. In the mouse, inhibiting the rate-limiting enzyme of polyamine synthesis, ornithine decarboxylase (ODC1) during early pregnancy largely prevents implantation, but the fate of the nonimplanted embryos is unknown. To determine whether polyamines control mouse embryonic diapause, we treated pregnant mice with an ODC1 inhibitor from d3.5 to d6.5 postcoitum. At d7.5, 72% of females had no signs of implantation whilst the remaining females exhibited disrupted placental formation and degenerate embryos. In the females with no implantation, we obtained viable blastocysts that had attenuated cell proliferation, indicating a state of diapause. When cultured in vitro, these exhibited trophoblast outgrowth, indicative of reactivation of embryogenesis. In contrast, direct culture of d3.5 blastocysts with an ODC1 inhibitor failed to cause entry into diapause. Examination of the polyamine pathway enzymes and a number of implantation factors indicated inhibition of ODC1 resulted in a uterine phenotype that resembled diapause, with some compensatory increases in crucial genes. Thus, we conclude that an absence or paucity of polyamines induces the uterine quiescence that causes entry of the blastocyst into embryonic diapause.
Sigma-Aldrich
M2 medium, With HEPES, without penicillin and streptomycin, liquid, sterile-filtered, suitable for mouse embryo cell culture
Pricing and availability is not currently available.