Merck
  • Membrane dynamics of resting and internalin B-bound MET receptor tyrosine kinase studied by single-molecule tracking.
Membrane dynamics of resting and internalin B-bound MET receptor tyrosine kinase studied by single-molecule tracking.
FEBS open bio (2017-09-15)
Marie-Lena I E Harwardt, Phoebe Young, Willem M Bleymüller, Timo Meyer, Christos Karathanasis, Hartmut H Niemann, Mike Heilemann, Marina S Dietz
ABSTRACT
The human MET receptor tyrosine kinase contributes to vertebrate development and cell proliferation. As a proto-oncogene, it is a target in cancer therapies. MET is also relevant for bacterial infection by
Sigma-Aldrich
Dulbecco′s Phosphate Buffered Saline, 10×, Modified, without calcium chloride and magnesium chloride, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Triton X-100, laboratory grade