Merck
Assessing the Risk for Resistance and Elucidating the Genetics of
Frontiers in microbiology (2017-10-04)
Can Zhang, Yongzhao Diao, Weizhen Wang, Jianjun Hao, Muhammad Imran, Hongxia Duan, Xili Liu
ABSTRACT
The genus
Product Description
Supelco
Myclobutanil, PESTANAL®, analytical standard
Supelco
Tebuconazole, PESTANAL®, analytical standard