Merck
  • Low-Temperature Wet Conformal Nickel Silicide Deposition for Transistor Technology through an Organometallic Approach.
Low-Temperature Wet Conformal Nickel Silicide Deposition for Transistor Technology through an Organometallic Approach.
ACS applied materials & interfaces (2017-01-13)
Tsung-Han Lin, Tigran Margossian, Michele De Marchi, Maxime Thammasack, Dmitry Zemlyanov, Sudhir Kumar, Jakub Jagielski, Li-Qing Zheng, Chih-Jen Shih, Renato Zenobi, Giovanni De Micheli, David Baudouin, Pierre-Emmanuel Gaillardon, Christophe Copéret
ABSTRACT
The race for performance of integrated circuits is nowadays facing a downscale limitation. To overpass this nanoscale limit, modern transistors with complex geometries have flourished, allowing higher performance and energy efficiency. Accompanying this breakthrough, challenges toward high-performance devices have emerged on each significant step, such as the inhomogeneous coverage issue and thermal-induced short circuit issue of metal silicide formation. In this respect, we developed a two-step organometallic approach for nickel silicide formation under near-ambient temperature. Transmission electron and atomic force microscopy show the formation of a homogeneous and conformal layer of NiSi