Merck
  • Parkin protects mitochondrial genome integrity and supports mitochondrial DNA repair.
Parkin protects mitochondrial genome integrity and supports mitochondrial DNA repair.
Human molecular genetics (2009-07-21)
Oliver Rothfuss, Heike Fischer, Takafumi Hasegawa, Martina Maisel, Petra Leitner, Franziska Miesel, Manu Sharma, Antje Bornemann, Daniela Berg, Thomas Gasser, Nadja Patenge
ABSTRACT
Mutations in the parkin gene are the most common cause of recessive familial Parkinson disease (PD). Parkin has been initially characterized as an ubiquitin E3 ligase, but the pathological relevance of this activity remains uncertain. Recently, an impressive amount of evidence has accumulated that parkin is involved in the maintenance of mitochondrial function and biogenesis. We used a human neuroblastoma cell line as a model to study the influence of endogenous parkin on mitochondrial genomic integrity. Using an unbiased chromatin immunoprecipitation approach, we found that parkin is associated physically with mitochondrial DNA (mtDNA) in proliferating as well as in differentiated SH-SY5Y cells. In vivo, the association of parkin with mtDNA could be confirmed in brain tissue of mouse and human origin. Replication and transcription of mtDNA were enhanced in SH-SY5Y cells over-expressing the parkin gene. The ability of parkin to support mtDNA-metabolism was impaired by pathogenic parkin point mutations. Most importantly, we show that parkin protects mtDNA from oxidative damage and stimulates mtDNA repair. Moreover, higher susceptibility of mtDNA to reactive oxygen species and reduced mtDNA repair capacity was observed in parkin-deleted fibroblasts of a PD patient. Our data indicate a novel role for parkin in directly supporting mitochondrial function and protecting mitochondrial genomic integrity from oxidative stress.
Sigma-Aldrich
Nuclease micrococcal from Staphylococcus aureus, 100-300 units/mg protein
Pricing and availability is not currently available.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Pricing and availability is not currently available.