Merck
  • miR-188 promotes senescence of lineage-negative bone marrow cells by targeting MAP3K3 expression.
miR-188 promotes senescence of lineage-negative bone marrow cells by targeting MAP3K3 expression.
FEBS letters (2017-06-24)
Yue Zheng, Hua Liu, Ye Kong
ABSTRACT
Lineage-negative bone marrow cells (lin-BMCs) have reparative potential for overcoming endothelial dysfunction and reducing cardiovascular risk. Here, we found that miR-188 is upregulated and mitogen-activated protein kinase kinase kinase 3 (MAP3K3) is downregulated in aged lin-BMCs, whereas their expression is reversed in young lin-BMCs. We identified and confirmed MAP3K3 as a direct target of miR-188. MiR-188 overexpression or MAP3K3 silencing in young lin-BMCs increases p16 and p21 expression, enhances cell senescence, and decreases the ability for cell proliferation, migration, and tube formation. Conversely, miR-188 suppression in aged lin-BMCs yields the opposite results. We further found that MAP3K3 is involved in miR-188-induced promotion of lin-BMC senescence. All data reveal that miR-188 induces lin-BMC senescence by targeting MAP3K3 expression, thus, providing new theoretical basis for the prevention and treatment of cardiovascular diseases.
Sigma-Aldrich
MISSION® esiRNA, targeting human MAP3K3
Pricing and availability is not currently available.