Merck
Effects of natural and synthetic isothiocyanate-based H
Neuropharmacology (2017-04-23)
Lorenzo Di Cesare Mannelli, Elena Lucarini, Laura Micheli, Ilaria Mosca, Paolo Ambrosino, Maria Virginia Soldovieri, Alma Martelli, Lara Testai, Maurizio Taglialatela, Vincenzo Calderone, Carla Ghelardini
ABSTRACT
Hydrogen sulfide (H
Sigma-Aldrich
GYY4137 Dichloromethane complex, ≥98% (HPLC)