- Manifestation of Geometric and Electronic Shell Structures of Metal Clusters in Intercluster Reactions.
Manifestation of Geometric and Electronic Shell Structures of Metal Clusters in Intercluster Reactions.
ACS nano (2017-05-18)
K R Krishnadas, Ananya Baksi, Atanu Ghosh, Ganapati Natarajan, Thalappil Pradeep
PMID28514137
ABSTRACT
Monolayer protected clusters exhibit rich diversity in geometric and electronic structures. However, structure-reactivity relationships in these clusters are rarely explored. In this context, [Ag