Merck
  • Interleukin-10-mediated regenerative postnatal tissue repair is dependent on regulation of hyaluronan metabolism
Interleukin-10-mediated regenerative postnatal tissue repair is dependent on regulation of hyaluronan metabolism
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2016-12-03)
Swathi Balaji, Xinyi Wang, Alice King, Louis D Le, Sukanta S Bhattacharya, Chad M Moles, Manish J Butte, Vinicio A de Jesus Perez, Kenneth W Liechty, Thomas N Wight, Timothy M Crombleholme, Paul L Bollyky, Sundeep G Keswani
ABSTRACT
The cytokine IL-10 has potent antifibrotic effects in models of adult fibrosis, but the mechanisms of action are unclear. Here, we report a novel finding that IL-10 triggers a signal transducer and activator of transcription 3 (STAT3)-dependent signaling pathway that regulates hyaluronan (HA) metabolism and drives adult fibroblasts to synthesize an HA-rich pericellular matrix, which mimics the fetal regenerative wound healing phenotype with reduced fibrosis. By using cre-lox-mediated novel, inducible, fibroblast-, keratinocyte-, and wound-specific STAT3-knockdown postnatal mice-plus syngeneic fibroblast cell-transplant models-we demonstrate that the regenerative effects of IL-10 in postnatal wounds are dependent on HA synthesis and fibroblast-specific STAT3-dependent signaling. The importance of IL-10-induced HA synthesis for regenerative wound healing is demonstrated by inhibition of HA synthesis in a murine wound model by administering 4-methylumbelliferone. Although IL-10 and STAT3 signaling were intact, the antifibrotic repair phenotype that is induced by IL-10 overexpression was abrogated in this model. Our data show a novel role for IL-10 beyond its accepted immune-regulatory mechanism. The opportunity for IL-10 to regulate a fibroblast-specific formation of a regenerative, HA-rich wound extracellular matrix may lead to the development of innovative therapies to attenuate postnatal fibrosis in organ systems or diseases in which dysregulated inflammation and HA intersect.-Balaji, S., Wang, X., King, A., Le, L. D., Bhattacharya, S. S., Moles, C. M., Butte, M. J., de Jesus Perez, V. A., Liechty, K. W., Wight, T. N., Crombleholme, T. M., Bollyky, P. L., Keswani, S. G. Interleukin-10-mediated regenerative postnatal tissue repair is dependent on regulation of hyaluronan metabolism
Sigma-Aldrich
Hyaluronidase from Streptomyces hyalurolyticus
Pricing and availability is not currently available.