- Thioredoxin A Is Essential for Motility and Contributes to Host Infection of
Thioredoxin A Is Essential for Motility and Contributes to Host Infection of
Frontiers in cellular and infection microbiology (2017-07-14)
Changyong Cheng, Zhimei Dong, Xiao Han, Hang Wang, Li Jiang, Jing Sun, Yongchun Yang, Tiantian Ma, Chunyan Shao, Xiaodu Wang, Zhongwei Chen, Weihuan Fang, Nancy E Freitag, Huarong Huang, Houhui Song
PMID28702378
ABSTRACT
Microbes employ the thioredoxin system to defend against oxidative stress and ensure correct disulfide bonding to maintain protein function.