Merck
  • A Novel Inhibitor IDPP Interferes with Entry and Egress of HCV by Targeting Glycoprotein E1 in a Genotype-Specific Manner.
A Novel Inhibitor IDPP Interferes with Entry and Egress of HCV by Targeting Glycoprotein E1 in a Genotype-Specific Manner.
Scientific reports (2017-03-24)
Myungeun Lee, Jaewon Yang, Eunji Jo, Ji-Young Lee, Hee-Young Kim, Ralf Bartenschlager, Eui-Cheol Shin, Yong-Soo Bae, Marc P Windisch
ABSTRACT
Despite recent advances in curing chronic hepatitis C (CHC), the high economic burden to therapy, viral drug resistance, difficult to treat hepatitis C virus (HCV) genotypes and patient groups are still of concern. To address this unmet medical needs, we devised strategies to identify novel viral interventions through target-free high-throughput screening of small molecules utilizing a phenotypic-based HCV infection assay. Thereby, a very potent (EC
Sigma-Aldrich
OptiPrep Density Gradient Medium, used for cell and subcellular organelle isolation
Pricing and availability is not currently available.
Sigma-Aldrich
2′-C-Methylcytidine, ≥95% (HPLC), powder
Pricing and availability is not currently available.
Sigma-Aldrich
Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)
Pricing and availability is not currently available.