- TOR-Dependent and -Independent Pathways Regulate Autophagy in
TOR-Dependent and -Independent Pathways Regulate Autophagy in
Frontiers in plant science (2017-07-27)
Yunting Pu, Xinjuan Luo, Diane C Bassham
PMID28744293
ABSTRACT
Autophagy is a critical process for recycling of cytoplasmic materials during environmental stress, senescence and cellular remodeling. It is upregulated under a wide range of abiotic stress conditions and is important for stress tolerance. Autophagy is repressed by the protein kinase target of rapamycin (TOR), which is activated in response to nutrients and in turn upregulates cell growth and translation and inhibits autophagy. Down-regulation of TOR in