Merck
  • Assessing the Intracellular Integrity of Phosphine-Stabilized Ultrasmall Cytotoxic Gold Nanoparticles Enabled by Fluorescence Labeling.
Assessing the Intracellular Integrity of Phosphine-Stabilized Ultrasmall Cytotoxic Gold Nanoparticles Enabled by Fluorescence Labeling.
Advanced healthcare materials (2016-11-20)
Janine Broda, Andrea Küster, Stefan Westhues, Dirk Fahrenkamp, Andreas T J Vogg, Julia Steitz, Felix M Mottaghy, Gerhard Müller-Newen, Ulrich Simon
ABSTRACT
As the size of nanoparticles (NPs) is in the range of biological molecules and subcellular structures, they provide new perspectives in biomedicine. This work presents studies concerning the cellular uptake and distribution of phosphine-stabilized cytotoxic 1.4 nm sized AuNPs and their probable degradation during this process. Therefore, ultrasmall phosphine-stabilized AuNPs are modified by linking a fluorophore covalently to the ligand shell. Monitoring the fluorescence on a cellular level by means of flow cytometry and confocal laser scanning microscopy allows determining the fate of the ligand shell during AuNP cell internalization, due to the fact that the fluorescence of a fluorophore bound near to the AuNP surface is quenched. Cell fractionation is conducted in order to quantify the AuNP content at the cell membrane, in the cytoplasm, and the cell nucleus. The incubation of cells with the fluorophore-modified AuNPs reveals a partial loss of the ligand shell upon AuNP cell interaction, evident by the emerging fluorescence signal. This loss is the precondition to unfold high AuNP cytotoxicity. Together with their significantly different biodistribution and enhanced circulation times compared to larger AuNPs, the findings demonstrate the high potential of ultrasmall AuNPs for drug development or therapy.
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide, ≥97.0% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
4-(Diphenylphosphino)benzoic acid, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
Pricing and availability is not currently available.