Merck
  • Expression of GPR17, a regulator of oligodendrocyte differentiation and maturation, in Nasu-Hakola disease brains.
Expression of GPR17, a regulator of oligodendrocyte differentiation and maturation, in Nasu-Hakola disease brains.
Intractable & rare diseases research (2017-03-31)
Jun-Ichi Satoh, Yoshihiro Kino, Motoaki Yanaizu, Youhei Tosaki, Kenji Sakai, Tusyoshi Ishida, Yuko Saito
ABSTRACT
The G protein-coupled receptor 17 (GPR17), a Gi-coupled GPCR, acts as an intrinsic timer of oligodendrocyte differentiation and myelination. The expression of GPR17 is upregulated during differentiation of oligodendrocyte precursor cells (OPCs) into premyelinating oligodendrocytes (preoligodendrocytes), whereas it is markedly downregulated during terminal maturation of myelinating oligodendrocytes. Nasu-Hakola disease (NHD) is a rare autosomal recessive disorder caused by a loss-of-function mutation of either
Sigma-Aldrich
Anti-GPR17 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.