Merck
  • Carnosine modulates glutamine synthetase expression in senescent astrocytes exposed to oxygen-glucose deprivation/recovery.
Carnosine modulates glutamine synthetase expression in senescent astrocytes exposed to oxygen-glucose deprivation/recovery.
Brain research bulletin (2017-01-25)
Xiaojie Shi, Bingyu Wang, Yuan Liu, Jingjing Zhang, Yuyan Huang, Pei Cao, Yao Shen, Jianxin Lyu
ABSTRACT
Carnosine is believed to be neuroprotective in cerebral ischemia. However, few reports concern its function on senescent astrocytes during cerebral ischemia. The aim of this study was to investigate the effects of carnosine on cell damage and glutamine synthetase (GS) expression in D-galactose-induced senescent astrocytes exposed to oxygen-glucose deprivation/recovery (OGD/R). The results showed that OGD/R caused massive cell damage and a significant decrease in GS expression both in the young and senescent astrocytes. The GS expression level was partly recovered whereas it continued to decline in the recovery stage in the young and senescent astrocytes, respectively. Decreased GS expression significantly inhibited glutamate uptake and glutamine production and release. Carnosine prevented the cell damage, rescued the expression of GS and reversed the glutamate uptake activity and glutamine production in the senescent astrocytes exposed to OGD/R. The modulatory effect of carnosine on GS expression was partly antagonized by pyrilamine, a selective histamine H
Sigma-Aldrich
Bisindolylmaleimide II, ≥97% (Mixture of 2 isomers)
Pricing and availability is not currently available.