Merck
  • Novel role for vascular endothelial growth factor (VEGF) receptor-1 and its ligand VEGF-B in motor neuron degeneration.
Novel role for vascular endothelial growth factor (VEGF) receptor-1 and its ligand VEGF-B in motor neuron degeneration.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2008-10-17)
Koen Poesen, Diether Lambrechts, Philip Van Damme, Joke Dhondt, Florian Bender, Nicolas Frank, Elke Bogaert, Bart Claes, Line Heylen, An Verheyen, Katrien Raes, Marc Tjwa, Ulf Eriksson, Masabumi Shibuya, Rony Nuydens, Ludo Van Den Bosch, Theo Meert, Rudi D'Hooge, Michael Sendtner, Wim Robberecht, Peter Carmeliet
ABSTRACT
Although vascular endothelial growth factor-B (VEGF-B) is a homolog of the angiogenic factor VEGF, it has only minimal angiogenic activity, raising the question of whether this factor has other (more relevant) biological properties. Intrigued by the possibility that VEGF family members affect neuronal cells, we explored whether VEGF-B might have a role in the nervous system. Here, we document that the 60 kDa VEGF-B isoform, VEGF-B(186), is a neuroprotective factor. VEGF-B(186) protected cultured primary motor neurons against degeneration. Mice lacking VEGF-B also developed a more severe form of motor neuron degeneration when intercrossed with mutant SOD1 mice. The in vitro and in vivo effects of VEGF-B(186) were dependent on the tyrosine kinase activities of its receptor, Flt1, in motor neurons. When delivered intracerebroventricularly, VEGF-B(186) prolonged the survival of mutant SOD1 rats. Compared with a similar dose of VEGF, VEGF-B(186) was safer and did not cause vessel growth or blood-brain barrier leakiness. The neuroprotective activity of VEGF-B, in combination with its negligible angiogenic/permeability activity, offers attractive opportunities for the treatment of neurodegenerative diseases.
Sigma-Aldrich
Mammalian Cell Lysis Kit