Merck
Synthesis of new photosensitive H
Dalton transactions (Cambridge, England : 2003) (2017-03-04)
M Stylianou, I Hadjiadamou, C Drouza, S C Hayes, E Lariou, I Tantis, P Lianos, A C Tsipis, A D Keramidas
ABSTRACT
A new two-electron photosensitizer, H
Sigma-Aldrich
Di-(2-picolyl)amine, 97%