Merck
  • Detection of endocrine disrupting chemicals and evidence of their effects on the HPG axis of the European anchovy Engraulis encrasicolus.
Detection of endocrine disrupting chemicals and evidence of their effects on the HPG axis of the European anchovy Engraulis encrasicolus.
Marine environmental research (2017-04-17)
Andrea Miccoli, Francesca Maradonna, Andrea De Felice, Vincenzo Caputo Barucchi, Andone Estonba, Michele Genangeli, Sauro Vittori, Iole Leonori, Oliana Carnevali
ABSTRACT
Natural/synthetic Endocrine Disrupting Chemicals (EDCs) may display estrogenic activity and a lower potency than 17β-estradiol. Nonetheless, their concentrations and additive effects can affect the endocrine system and reproductive processes related to the Hypothalamic-Pituitary-Gonadal (HPG) axis. Because of their persistence in both the environment and biological systems, they ultimately target multi-level predators, including humans. We detected presence and effects of xenobiotics on wild anchovy Engraulis encrasicolus in the Western Adriatic Sea. Twenty-one PCBs and five organochlorines were detected on the order of ng g