Merck
  • Synthesis and characterisation of non-bonded 1.7μm thin-shell (TS1.7-100nm) silica particles for the rapid separation and analysis of uric acid and creatinine in human urine by hydrophilic interaction chromatography.
Synthesis and characterisation of non-bonded 1.7μm thin-shell (TS1.7-100nm) silica particles for the rapid separation and analysis of uric acid and creatinine in human urine by hydrophilic interaction chromatography.
Journal of chromatography. A (2017-05-27)
Victor K Langsi, Benjamin A Ashu-Arrah, Niamh Ward, Jeremy D Glennon
ABSTRACT
A rapid chromatographic method for the simultaneous determination of uric acid (UA) and creatinine (Cr) in human urine is described, using a non-bonded 1.7μm thin-shell (TS1.7-100nm) silica particle prepared by the seeded-growth approach. The new shell particle was characterised by scanning electron microscopy (SEM), transmission electron microscopy (TEM), dynamic light scattering (DLS) and BET analysis. TEM reveals 1.5μm solid core and 100nm shell thickness. DLS shows polydispersity index <0.2. Expanded pore size of 88Å and specific surface area of 78m
Sigma-Aldrich
Creatinine hydrochloride, ≥97%
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium hydroxide solution, puriss., 30-33% NH3 in H2O
Pricing and availability is not currently available.