Merck
  • Selective Activation of Estrogen Receptor α Activation Function-1 Is Sufficient to Prevent Obesity, Steatosis, and Insulin Resistance in Mouse.
Selective Activation of Estrogen Receptor α Activation Function-1 Is Sufficient to Prevent Obesity, Steatosis, and Insulin Resistance in Mouse.
The American journal of pathology (2017-05-16)
Maeva Guillaume, Sandra Handgraaf, Aurélie Fabre, Isabelle Raymond-Letron, Elodie Riant, Alexandra Montagner, Alexia Vinel, Melissa Buscato, Natalia Smirnova, Coralie Fontaine, Hervé Guillou, Jean-François Arnal, Pierre Gourdy
ABSTRACT
Estrogen receptor α (ERα) regulates gene transcription through two activation functions (ERα-AF1 and ERα-AF2). We recently found that the protection conferred by 17β-estradiol against obesity and insulin resistance requires ERα-AF2 but not ERα-AF1. However, the interplay between the two ERα-AFs is poorly understood in vivo and the metabolic influence of a specific ERα-AF1 action remains to be explored. To this end, wild-type, ERα-deficient, or ERα-AF1-deficient ovariectomized female mice were fed a high-fat diet and concomitantly administered with vehicle or tamoxifen, a selective ER modulator that acts as a ERα-AF1 agonist/ERα-AF2 antagonist. In ovariectomized wild-type mice, tamoxifen significantly reduced food intake and totally prevented adiposity, insulin resistance, and steatosis. These effects were abolished in ERα-deficient and ERα-AF1-deficient mice, revealing the specific role of ERα-AF1 activation. Finally, hepatic gene expression changes elicited by tamoxifen in wild-type mice were abrogated in ERα-AF1-deficient mice. The combination of pharmacologic and transgenic approaches thus indicates that selective ERα-AF1 activation by tamoxifen is sufficient to elicit metabolic protection, contrasting with the specific requirement of ERα-AF2 in the metabolic actions of 17β-estradiol. This redundancy in the ability of the two ERα-AFs to separately mediate metabolic prevention strikingly contrasts with the contribution of both ERα-AFs in breast cancer proliferation, shedding new light on the therapeutic potential of selective ER modulation.
Product Description
Millipore
MILLIPLEX® Mouse Adiponectin Magnetic Bead- Single Plex - Endocrine Assay, This MILLIPLEX MAP Mouse Adiponectin Magnetic Bead Single Plex Kit is an Endocrine single plex assay. The analyte available for this kit is Adiponectin.