- Alloy Acceptor: Superior Alternative to PCBM toward Efficient and Stable Organic Solar Cells.
Alloy Acceptor: Superior Alternative to PCBM toward Efficient and Stable Organic Solar Cells.
Advanced materials (Deerfield Beach, Fla.) (2016-06-24)
Pei Cheng, Cenqi Yan, Yang Wu, Jiayu Wang, Meng Qin, Qiaoshi An, Jiamin Cao, Lijun Huo, Fujun Zhang, Liming Ding, Yanming Sun, Wei Ma, Xiaowei Zhan
PMID27337385
ABSTRACT
The alloy acceptor (indene-C