Merck
  • One single, fast and robust capillary electrophoresis method for the direct quantification of intact adenovirus particles in upstream and downstream processing samples.
One single, fast and robust capillary electrophoresis method for the direct quantification of intact adenovirus particles in upstream and downstream processing samples.
Talanta (2017-02-19)
Ewoud van Tricht, Lars Geurink, Harold Backus, Marta Germano, Govert W Somsen, Cari E Sänger-van de Griend
ABSTRACT
During development of adenovirus-based vaccines, samples have to be analyzed in order to either monitor the production process or control the quality and safety of the product. An important quality attribute is the total concentration of intact adenoviruses, which currently is determined by quantitative polymerase chain reaction (qPCR) or anion exchange-HPLC. Capillary Electrophoresis (CE) was evaluated as alternative to the current methods with the aim to have one single method that allows reliable and fast quantification of adenovirus particles throughout the full process. Intact adenoviruses samples from downstream processing and upstream processing were analyzed directly by CE with UV-detection at 214nm. Only the samples with high amounts of DNA required a simple sample pretreatment by benzonase. Adenovirus particles were separated from matrix components such as cell debris, residual cell DNA, and/or proteins on a PVA-coated capillary using a BGE consisting of 125mM Tris, 338mM tricine and 0.2% v/v polysorbate-20 at pH 7.7. Full factorial design of experiments was used for method optimization as part of the analytical quality by design (AQbD) method development approach. The method was validated for the quantification of adenoviruses on five representative samples from the manufacturing process in the range of 0.5×10
Sigma-Aldrich
Dextran sulfate sodium salt from Leuconostoc spp., for molecular biology, average Mw >500,000 (dextran starting material), contains 0.5-2% phosphate buffer
Pricing and availability is not currently available.