Merck
  • Characterization and source profiling of volatile organic compounds in indoor air of private residences in Selangor State, Malaysia.
Characterization and source profiling of volatile organic compounds in indoor air of private residences in Selangor State, Malaysia.
The Science of the total environment (2017-02-27)
Nobumitsu Sakai, Shuta Yamamoto, Yasuto Matsui, Md Firoz Khan, Mohd Talib Latif, Mustafa Ali Mohd, Minoru Yoneda
ABSTRACT
Volatile Organic Compounds (VOCs) in indoor air were investigated at 39 private residences in Selangor State, Malaysia to characterize the indoor air quality and to identify pollution sources. Twenty-two VOCs including isomers (14 aldehydes, 5 aromatic hydrocarbons, acetone, trichloroethylene and tetrachloroethylene) were collected by 2 passive samplers for 24h and quantitated using high performance liquid chromatography and gas chromatography mass spectrometry. Source profiling based on benzene/toluene ratio as well as statistical analysis (cluster analysis, bivariate correlation analysis and principal component analysis) was performed to identify pollution sources of the detected VOCs. The VOCs concentrations were compared with regulatory limits of air quality guidelines in WHO/EU, the US, Canada and Japan to clarify the potential health risks to the residents. The 39 residences were classified into 2 groups and 2 ungrouped residences based on the dendrogram in the cluster analysis. Group 1 (n=30) had mainly toluene (6.87±2.19μg/m
Sigma-Aldrich
2,5-Dimethylbenzaldehyde, 99%
Pricing and availability is not currently available.