Merck
  • TRPA1, substance P, histamine and 5-hydroxytryptamine interact in an interdependent way to induce nociception.
TRPA1, substance P, histamine and 5-hydroxytryptamine interact in an interdependent way to induce nociception.
Inflammation research : official journal of the European Histamine Research Society ... [et al.] (2016-12-03)
Luana Fischer, Maria Isabel Lavoranti, Mariana de Oliveira Borges, Alana Farias Miksza, Natalia Fantin Sardi, Bruno Jacson Martynhak, Claudia H Tambeli, Carlos Amílcar Parada
ABSTRACT
Although TRPA1, SP, histamine and 5-hydroxytryptamine (5-HT) have recognized contribution to nociceptive mechanisms, little is known about how they interact with each other to mediate inflammatory pain in vivo. In this study we evaluated whether TRPA1, SP, histamine and 5-HT interact, in an interdependent way, to induce nociception in vivo. The subcutaneous injection of the TRPA1 agonist allyl isothiocyanate (AITC) into the rat's hind paw induced a dose-dependent and short lasting behavioral nociceptive response that was blocked by the co-administration of the TRPA1 antagonist, HC030031, or by the pretreatment with antisense ODN against TRPA1. AITC-induced nociception was significantly decreased by the co-administration of selective antagonists for the NK1 receptor for substance P, the H1 receptor for histamine and the 5-HT Together, these findings suggest a self-sustainable cycle around TRPA1, no matter where the cycle is initiated each step is achieved and even subeffective activation of more than one step results in a synergistic activation of the overall cycle.
Sigma-Aldrich
HC-030031, ≥98% (HPLC), powder
Pricing and availability is not currently available.