Merck
  • The Plant Hormone Abscisic Acid Is a Prosurvival Factor in Human and Murine Megakaryocytes.
The Plant Hormone Abscisic Acid Is a Prosurvival Factor in Human and Murine Megakaryocytes.
The Journal of biological chemistry (2017-01-05)
Alessandro Malara, Chiara Fresia, Christian Andrea Di Buduo, Paolo Maria Soprano, Francesco Moccia, Cesare Balduini, Elena Zocchi, Antonio De Flora, Alessandra Balduini
ABSTRACT
Abscisic acid (ABA) is a phytohormone involved in pivotal physiological functions in higher plants. Recently, ABA has been proven to be also secreted and active in mammals, where it stimulates the activity of innate immune cells, mesenchymal and hematopoietic stem cells, and insulin-releasing pancreatic β cells through a signaling pathway involving the second messenger cyclic ADP-ribose (cADPR). In addition to behaving like an animal hormone, ABA also holds promise as a nutraceutical plant-derived compound in humans. Many biological functions of ABA in mammals are mediated by its binding to the LANCL-2 receptor protein. A putative binding of ABA to GRP78, a key regulator of endoplasmic reticulum stress, has also been proposed. Here we investigated the role of exogenous ABA in modulating thrombopoiesis, the process of platelet generation. Our results demonstrate that expression of both LANCL-2 and GRP78 is up-regulated during hematopoietic stem cell differentiation into mature megakaryocytes (Mks). Functional ABA receptors exist in mature Mks because ABA induces an intracellular Ca
Sigma-Aldrich
Econazole nitrate salt
Pricing and availability is not currently available.