MilliporeSigma
  • Overexpression of the Endosomal Anion/Proton Exchanger ClC-5 Increases Cell Susceptibility toward
Overexpression of the Endosomal Anion/Proton Exchanger ClC-5 Increases Cell Susceptibility toward
Frontiers in cellular and infection microbiology (2017-03-30)
Frederike Ruhe, Alexandra Olling, Rasmus Abromeit, Dennis Rataj, Matthias Grieschat, Andre Zeug, Ralf Gerhard, Alexi Alekov
ABSTRACT
Virulent
Sigma-Aldrich
MISSION® esiRNA, targeting human CLCN5