Merck
Highly Emissive Covalent Organic Frameworks.
Journal of the American Chemical Society (2016-04-26)
Sasanka Dalapati, Enquan Jin, Matthew Addicoat, Thomas Heine, Donglin Jiang
ABSTRACT
Highly luminescent covalent organic frameworks (COFs) are rarely achieved because of the aggregation-caused quenching (ACQ) of π-π stacked layers. Here, we report a general strategy to design highly emissive COFs by introducing an aggregation-induced emission (AIE) mechanism. The integration of AIE-active units into the polygon vertices yields crystalline porous COFs with periodic π-stacked columnar AIE arrays. These columnar AIE π-arrays dominate the luminescence of the COFs, achieve exceptional quantum yield via a synergistic structural locking effect of intralayer covalent bonding and interlayer noncovalent π-π interactions and serve as a highly sensitive sensor to report ammonia down to sub ppm level. Our strategy breaks through the ACQ-based mechanistic limitations of COFs and opens a way to explore highly emissive COF materials.
Sigma-Aldrich
1,1,2,2-Tetrakis(4-bromophenyl)ethylene
Pricing and availability is not currently available.