Merck
  • The effects of baicalein on gastric mucosal ulcerations in mice: Protective pathways and anti-secretory mechanisms.
The effects of baicalein on gastric mucosal ulcerations in mice: Protective pathways and anti-secretory mechanisms.
Chemico-biological interactions (2016-10-27)
Ana Roseli S Ribeiro, José Diego do Nascimento Valença, Jeferson da Silva Santos, Thaise Boeing, Luisa Mota da Silva, Sérgio Faloni de Andrade, Ricardo L C Albuquerque-Júnior, Sara Maria Thomazzi
ABSTRACT
Many flavonoids have been shown to present good results for the treatment of gastric ulcers. Baicalein, a bioactive flavonoid derived from the Scutellaria baicalensis Georgi root, possesses several biological effects, such as anti-inflammatory and antioxidant. This study was conducted to assess the gastroprotective properties of baicalein. Anti-ulcerogenic assay was performed using the protocol of ulcer induced by ethanol/HCl in mice; then, the role of presynaptic α2-receptors, sulfhydryl (SH) compounds, nitric oxide (NO), prostaglandin (PG) and ATP-sensitive K+ (KATP) channels in gastroprotection of baicalein was investigated. The levels of reduced glutathione (GSH) and the myeloperoxidase (MPO) activity were measured in the gastric mucosa. Parameters of gastric secretion (volume, [H+] and pH) were determined with or without the presence of the secretagogue agent histamine, as well as mucus in gastric contents, by the pylorus ligation model. In vitro H+,K+-ATPase activity was also determined. Baicalein (10, 30 and 100 mg/kg) exhibited a dose related gastroprotective effect (P < 0.001) against acidified ethanol-induced lesions. The intraperitoneal treatment of mice with a α2-adrenoreceptor antagonist (yohimbine; 2 mg/kg), a SH compounds blocker (N-ethylmaleimide, NEM; 10 mg/kg), a non-selective inhibitor of NO synthase (Nw-nitro-L-arginine methyl ester hydrochloride, L-NAME; 10 mg/kg), a non-selective inhibitor of cyclo-oxygenase (indomethacin; 10 mg/kg) or a KATP channel blocker (glibenclamide; 10 mg/kg) was able to reverse (P < 0.001) the gastroprotective response caused by baicalein (30 mg/kg). Baicalein (30 mg/kg; P < 0.05) was able to increase GSH levels and decreasing MPO activity. The intraduodenal treatment with baicalein (30 and 100 mg/kg) significantly increased (P < 0.05) the gastric mucus secretion. Additionally, the treatment with baicalein reduced (30 and 100 mg/kg; P < 0.05) the secretion volume and total acid secretion, and also increased (10, 30 and 100 mg/kg; P < 0.001) the pH value, after pylorus ligature. Baicalein (30 mg/kg) was also effective in inhibiting the effects of histamine on gastric secretion (volume, [H+] and pH; P < 0.001). Baicalein at 10 and 30 μg/mL showed anti-H+,K+-ATPase activity. In conclusion, the present results provide convincing evidence that baicalein could be used as a cytoprotective (preventive effect) and anti-ulcerogenic (anti-secretory effect) agent in the gastric ulcers.
Sigma-Aldrich
Baicalein, 98%
Pricing and availability is not currently available.
Ouabain, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.