Merck
  • HMGB1-mediated autophagy decreases sensitivity to oxymatrine in SW982 human synovial sarcoma cells.
HMGB1-mediated autophagy decreases sensitivity to oxymatrine in SW982 human synovial sarcoma cells.
Scientific reports (2016-11-30)
Yongsong Cai, Peng Xu, Le Yang, Ke Xu, Jialin Zhu, Xiaoqing Wu, Congshan Jiang, Qiling Yuan, Bo Wang, Yuanbo Li, Yusheng Qiu
ABSTRACT
Oxymatrine (OMT) is a type of alkaloid extracted from a traditional Chinese medicinal herb, Sophora flavescens. Although the antitumor activities of OMT have been observed in various cancers, there are no reports regarding the effects of OMT on human synovial sarcoma. In the present study, we analyzed the antitumor activities of OMT in SW982 human synovial sarcoma cells and determine whether high mobility group box protein 1 (HMGB1)-mediated autophagy was associated with its therapeutic effects. We found that OMT exhibited antitumor activity in SW982 cells and facilitated increases in autophagy. Inhibition of autophagy by 3-MA or ATG7 siRNA increased the level of apoptosis, which indicated that OMT-induced autophagy protected cells from the cytotoxicity of OMT. Administration of OMT to SW982 cells increased the expression of HMGB1. When HMGB1 was inhibited via HMGB1-siRNA, OMT-induced autophagy was decreased, and apoptosis was increased. Furthermore, we found that HMGB1-siRNA significantly increased the expression of p-Akt and p-mTOR. OMT-induced autophagy may be mediated by the Akt/mTOR pathway, and HMGB1 plays a vital role in the regulation of autophagy. Therefore, we believe that combining OMT with an inhibitor of autophagy or HMGB1 may make OMT more effective in the treatment of human synovial sarcoma.
Sigma-Aldrich
MISSION® esiRNA, targeting human ATG7
Pricing and availability is not currently available.
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Pricing and availability is not currently available.