Merck
  • Local and systemic hormonal responses in pepper leaves during compatible and incompatible pepper-tobamovirus interactions.
Local and systemic hormonal responses in pepper leaves during compatible and incompatible pepper-tobamovirus interactions.
Plant physiology and biochemistry : PPB (2016-11-05)
Michał Dziurka, Anna Janeczko, Csilla Juhász, Gábor Gullner, Jana Oklestková, Ondrej Novák, Diana Saja, Andrzej Skoczowski, István Tóbiás, Balázs Barna
ABSTRACT
Phytohormone levels and the expression of genes encoding key enzymes participating in hormone biosynthetic pathways were investigated in pepper leaves inoculated with two different tobamoviruses. Obuda pepper virus (ObPV) inoculation led to the development of hypersensitive reaction (incompatible interaction), while Pepper mild mottle virus (PMMoV) inoculation resulted in a systemic, compatible interaction. ObPV-inoculation markedly increased not only the levels of salicylic acid (SA) (73-fold) and jasmonic acid (8-fold) but also those of abscisic acid, indole-3-acetic acid, indole-3-butyric acid, cis-zeatin, cis-zeatin-9-riboside and trans-zeatin-9-riboside in the inoculated pepper leaves 3 days post inoculation. PMMoV infection increased only the contents of gibberellic acid and SA. Hormone contents did not change significantly after ObPV or PMMoV infection in non-infected upper leaves 20 days post inoculation. Concentrations of some brassinosteroids (BRs) and progesterone increased both in ObPV- and PMMoV inoculated leaves. ObPV inoculation markedly induced the expression of three phenylalanine ammonia-lyase (PAL) and a 1-aminocyclopropane-1-carboxylate oxidase (ACO) genes, while that of an isochorismate synthase (ICS) gene was not modified. PMMoV inoculation did not alter the expression of PAL and ICS genes but induced the transcript abundance of ACO although later than ObPV. Pre-treatment of pepper leaves with exogenous 24-epi-brassinolide (24-epi-BR) prior to ObPV-inoculation strongly mitigated the visible symptoms caused by ObPV. In addition, 24-epi-BR pre-treatment markedly altered the level of several hormones in pepper leaves following ObPV-inoculation. These data indicate that ObPV- and PMMoV-inoculations lead to intricate but well harmonized hormonal responses that are largely determined by the incompatible or compatible nature of plant-virus interactions.
Sigma-Aldrich
Indole-3-butyric acid potassium salt, BioReagent, suitable for plant cell culture
Sigma-Aldrich
(+)-Abscisic acid, ≥98% (HPLC)