Merck
  • Low-level vagus nerve stimulation upregulates small conductance calcium-activated potassium channels in the stellate ganglion.
Low-level vagus nerve stimulation upregulates small conductance calcium-activated potassium channels in the stellate ganglion.
Heart rhythm (2013-01-30)
Mark J Shen, Hao-Che Chang, Hyung-Wook Park, A George Akingba, Po-Cheng Chang, Zheng Zhang, Shien-Fong Lin, Changyu Shen, Lan S Chen, Zhenhui Chen, Michael C Fishbein, Nipavan Chiamvimonvat, Peng-Sheng Chen
ABSTRACT
Small conductance calcium-activated potassium (SK) channels are responsible for afterhyperpolarization that suppresses nerve discharges. To test the hypothesis that low-level vagus nerve stimulation (LL-VNS) leads to the upregulation of SK2 proteins in the left stellate ganglion. Six dogs (group 1) underwent 1-week LL-VNS of the left cervical vagus nerve. Five normal dogs (group 2) were used as controls. SK2 protein levels were examined by using Western blotting. The ratio between SK2 and glyceraldehydes-3-phosphate-dehydrogenase levels was used as an arbitrary unit (AU). We found higher SK2 expression in group 1 (0.124 ± 0.049 AU) than in group 2 (0.085 ± 0.031 AU; P<.05). Immunostaining showed that the density of nerve structures stained with SK2 antibody was also higher in group 1 (11,546 ± 7,271 μm(2)/mm(2)) than in group 2 (5321 ± 3164 μm(2)/mm(2); P<.05). There were significantly more ganglion cells without immunoreactivity to tyrosine hydroxylase (TH) in group 1 (11.4%±2.3%) than in group 2 (4.9% ± 0.7%; P<.05). The TH-negative ganglion cells mostly stained positive for choline acetyltransferase (95.9% ± 2.8% in group 1 and 86.1% ± 4.4% in group 2; P = .10). Immunofluorescence confocal microscopy revealed a significant decrease in the SK2 staining in the cytosol but an increase in the SK2 staining on the membrane of the ganglion cells in group 1 compared to group 2. Left LL-VNS results in the upregulation of SK2 proteins, increased SK2 protein expression in the cell membrane, and increased TH-negative (mostly choline acetyltransferase-positive) ganglion cells in the left stellate ganglion. These changes may underlie the antiarrhythmic efficacy of LL-VNS in ambulatory dogs.
Sigma-Aldrich
Anti-Potassium Channel SK2 antibody produced in rabbit, affinity isolated antibody, lyophilized powder